missing translation for 'onlineSavingsMsg'
Learn More

Diethylaminosulfur trifluoride, 95%, Thermo Scientific Chemicals

Catalog Number 11481890
Click to view available options
Quantity:
5 g
25 g
100 g
38078-09-0
C4H10F3NS
161.186
MFCD00000363
CSJLBAMHHLJAAS-UHFFFAOYSA-N
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
123472
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
CCN(CC)S(F)(F)F
38078-09-0
C4H10F3NS
161.186
MFCD00000363
CSJLBAMHHLJAAS-UHFFFAOYSA-N
diethylaminosulfur trifluoride, dast, diethylaminosulphur trifluoride, diethylamino sulfur trifluoride, diethylaminosulfurtrifluoride, diethylaminosulphurtrifluoride, diethylamino sulphur trifluoride, unii-78622bv6ij, sulfur, n-ethylethanaminato trifluoro-, t-4
123472
N-ethyl-N-(trifluoro-$l^{4}-sulfanyl)ethanamine
CCN(CC)S(F)(F)F

Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylaminosulfur trifluoride (DAST) is used as a precursor to prepare 2,6-dimethoxybenzoyl fluoride and 2-thiazolines. It is a useful fluorinating agent used for a variety of compounds including alcohols, thioethers, alkenols and cyanohydrins. It also serves as a catalyst for Friedel-Crafts allylation reaction using tertiary cyclopropyl silyl ethers and in the rearrangement of homoallylic alcohols to unsaturated aldehydes. It plays an important role for gem difluorination of ketopipecolinic acids.

Solubility
Miscible with water, ethanol, benzene and acetonitrile.

Notes
Store in cool place. Moisture sensitive. It reacts with water and ethanol. Incompatible with water, acids and bases.
TRUSTED_SUSTAINABILITY
Density 1.23
Boiling Point 30°C to 32°C (3 mmHg)
Flash Point 23°C (73°F)
Linear Formula (CH3CH2)2NSF3
Refractive Index 1.4115
Quantity 5 g
UN Number UN2920
Beilstein 1849066
Sensitivity Moisture sensitive
Solubility Information Miscible with water,ethanol,benzene and acetonitrile.
Formula Weight 161.19
Percent Purity 95%
Chemical Name or Material Diethylaminosulfur trifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'