missing translation for 'onlineSavingsMsg'
Learn More

Diethylaminoethyl dextran

Product Code. 15465949
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465949 100 g 100g
15455949 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465949 Supplier Thermo Scientific Alfa Aesar Supplier No. J63781.22
  • ONLINE EXCLUSIVE PRICE
    £182.58
    £138.65 / 100g
    Save £43.93
    24% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9015-73-0

DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems. The use of DEAE-Dextran as an efficient adjuvant in vaccine production is well established. For instance, DEAE-Dextran has been used for immunization of guinea pigs and swine with inactivated foot-and-mouth disease and rhesus monkeys with inactivated Venezulean equine encephalomyelitis virus, IVEE. The mechanism of action of DEAE-Dextran is not clear. The effect on the humoral response of rhesus monkeys to IVEE vaccine results in a typical IgM-IgG response. It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9015-73-0
MDL Number MFCD00081561
Synonym DEAE-Dextran
IUPAC Name Diethylaminoethyl dextran

Specifications

Color White
Quantity 100 g
Merck Index 14,2936
Solubility Information Soluble in water.
Physical Form Powder
Chemical Name or Material Diethylaminoethyl dextran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.