missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, Mikromol™

Product Code. 17988879
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988879 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17988879 Supplier Mikromol Supplier No. MM0273.13
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Diethylamine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylamine
CAS 109-89-7
Analyte or Component Names Diethylamine
Shipping Condition Room Temperature
Molecular Formula C4 H11 N
Certifications/Compliance ISO 9001
Synonym N-Ethylethanamine, Diethylamine (8CI), DEA, N,N-Diethylamine, Melphalan Imp. K (EP), Melphalan Impurity K, Potassium Clavulanate Impurity I
InChI Formula InChI=1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3
SMILES CCNCC
IUPAC Name N-ethylethanamine
Molecular Weight (g/mol) 73.14
Quantity 100 mg
Formula Weight 73.0891 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.