missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 10387850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
1kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387850 1 kg Plastic bottle 1kg
10592551 100 g Plastic bottle 100g
10001720 500 g Plastic bottle 500g
13458378 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10387850 Supplier Thermo Scientific Acros Supplier No. 155890010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 660-68-4
MDL Number MFCD00012499
InChI Key HDITUCONWLWUJR-UHFFFAOYSA-N
Synonym diethylamine hydrochloride, diethylammonium chloride, diethyl amine hydrochloride, n-ethylethanamine hydrochloride, ethanamine, n-ethyl-, hydrochloride, unii-ze9v3g1135, ethanamine, n-ethyl-, hydrochloride 1:1, diethylaminehydrochloride, diethylamine hcl, diethyl amine hcl
PubChem CID 10197650
IUPAC Name N-ethylethanamine;hydrochloride
SMILES CCNCC.Cl

Specifications

Melting Point 227.0°C to 230.0°C
Color White
Boiling Point 320.0°C to 330.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Molecular Formula C4H12ClN
Linear Formula (C2H5)2NH·HCl
Quantity 1 kg
Molecular Weight (g/mol) 109.597
Formula Weight 109.6
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Diethylamine hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Diethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.