missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethylamine hydrobromide, 98%

Product Code. 11408673
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408673 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408673 Supplier Alfa Aesar™ Supplier No. A10452.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 6274-12-0
Melting Point 218°C to 220°C
Molecular Formula C4H12BrN
MDL Number MFCD00012498
Quantity 50 g
Beilstein 3905681
Synonym diethylamine hydrobromide, ethanamine, n-ethyl-, hydrobromide, diethylamine hbr, acmc-1b9lr, n-ethylethanamine hydrobromide
InChI Key AATGHKSFEUVOPF-UHFFFAOYSA-N
SMILES [Br-].CC[NH2+]CC
IUPAC Name diethylazanium bromide
Molecular Weight (g/mol) 154.05
PubChem CID 10986453
Formula Weight 154.06
Percent Purity 98%
Chemical Name or Material Diethylamine hydrobromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.