missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylamine ≥ 99.5%, ChromAR™ for HPLC, Macron Fine Chemicals™
Description
- Ultraviolet Absorbance (1.00-cm cell vs water):
- at 300 nm max. 0.3
- at 320 nm max. 0.15
- at 350 nm max. 0.05
- at 400 nm max. 0.004
- Ultraviolet Absorbance (1.00-cm path vs water):
- at 400 nm max. 0.004
Specifications
Specifications
| CAS | 109-89-7 |
| Boiling Point | 56°C |
| Flash Point | -23°C |
| Assay Percent Range | min. 99.5% |
| Packaging | Glass bottle |
| Molecular Formula | C4H11N |
| MDL Number | MFCD00009032 |
| Quantity | 2.5 L |
| UN Number | 1486 |
| Residue after Evaporation | max. 0.005% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privatlivspolitik.
Ser du en mulighed for forbedring?