missing translation for 'onlineSavingsMsg'
Learn More

Diethylaluminum ethoxide, 25% w/w in hexane, Thermo Scientific™

Product Code. 11336329
Change view
Click to view available options
Quantity:
50g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336329 50g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11336329 Supplier Thermo Scientific Alfa Aesar Supplier No. 087625.NE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1586-92-1
Density 0.851 g/mL
Flash Point −23°C (−9°F)
Odor Petrol-like
Molecular Formula C6H15AlO
Linear Formula (C2H5)2Al(OC2H5)
MDL Number MFCD00009059
Quantity 50g
UN Number UN3394
Sensitivity Air and moisture sensitive
Synonym diethylaluminum ethoxide, aluminum, ethoxydiethyl, ethoxydiethylaluminium, diethylaluminium ethoxide, ethoxydiethylaluminum, ethoxy-diethylaluminum, ethoxydiethyl aluminum, ethoxy-diethylaluminium, ethoxydiethyl aluminium, ethoxydiethyl-aluminium
Solubility Information Soluble in hydrocarbons and ether.
InChI Key GCPCLEKQVMKXJM-UHFFFAOYSA-N
SMILES CCO[Al](CC)CC
IUPAC Name ethoxy(diethyl)alumane
Molecular Weight (g/mol) 130.17
PubChem CID 16683109
Formula Weight 130.17
Chemical Name or Material Diethylaluminum ethoxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.