missing translation for 'onlineSavingsMsg'
Learn More

Diethyl trichloromethylphosphonate, 98%

Product Code. p-7048371
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361398 2 g 2g
11371398 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361398 Supplier Thermo Scientific Alfa Aesar Supplier No. 010306.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 866-23-9 | C5H10Cl3O3P | 255.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866-23-9
Molecular Formula C5H10Cl3O3P
Molecular Weight (g/mol) 255.46
MDL Number MFCD00013666
InChI Key RVAQSYWDOSHWGP-UHFFFAOYSA-N
Synonym diethyl trichloromethylphosphonate, diethyl trichloromethyl phosphonate, phosphonic acid, trichloromethyl-, diethyl ester, methanephosphonic acid, trichloro-, diethyl ester, diethyltrichloromethylphosphonate, wln: gxggpo&o2&o2, 4-03-00-00262 beilstein handbook reference, trichloromethylphosphonic acid diethyl, diethoxyphosphino trichloromethyl-1-one
PubChem CID 70085
IUPAC Name 1-[ethoxy(trichloromethyl)phosphoryl]oxyethane
SMILES CCOP(=O)(OCC)C(Cl)(Cl)Cl

Specifications

Density 1.3670 g/mL
Boiling Point 128°C (14 mmHg)
Refractive Index 1.462
Quantity 2 g
UN Number UN3278
Formula Weight 255.48
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl trichloromethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.