missing translation for 'onlineSavingsMsg'
Learn More

Diethyl tetrafluorosuccinate, 97%

Product Code. 11379175
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379175 1 g 1g
11389175 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379175 Supplier Thermo Scientific Alfa Aesar Supplier No. L16469.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 377-71-9 | C8H10F4O4 | 246.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 377-71-9
Molecular Formula C8H10F4O4
Molecular Weight (g/mol) 246.158
MDL Number MFCD00015155
InChI Key BLZSSBPZHBFNBN-UHFFFAOYSA-N
Synonym diethyl tetrafluorosuccinate, diethyl perfluorosuccinate, butanedioic acid, tetrafluoro-, diethyl ester, diethyl 2,2,3,3-tetrafluorosuccinate, butanedioic acid, 2,2,3,3-tetrafluoro-, 1,4-diethyl ester, 1,4-diethyl 2,2,3,3-tetrafluorobutanedioate, diethyl 2,2,3,3-tetrafluorobutane-1,4-dioate, diethyltetrafluorosuccinate, ethyl perfluorosuccinate, acmc-1cqkj
PubChem CID 67834
IUPAC Name diethyl 2,2,3,3-tetrafluorobutanedioate
SMILES CCOC(=O)C(C(C(=O)OCC)(F)F)(F)F

Specifications

Density 1.273
Boiling Point 78°C (5 mmHg)
Refractive Index 1.368
Quantity 1 g
Beilstein 1798175
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material Diethyl tetrafluorosuccinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.