missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Terephthalate, TRC

Product Code. 30385335
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385335 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385335 Supplier Toronto Research Chemicals Supplier No. TRCD4451305G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Terephthalic Acid Diethyl Ester
CAS 636-09-9
Molecular Formula C12 H14 O4
Quantity 5 g
Synonym 1,4-Benzenedicarboxylic acid, 1,4-diethyl ester, 1,4-Benzenedicarboxylic acid, diethyl ester (9CI), Terephthalic acid, diethyl ester (6CI,7CI,8CI), Diethyl terephthalate, NSC 68816, p-Benzenedicarboxylic acid diethyl ester, p-Diethyl phthalate, Terephthalic acid diethyl ester, Terephthalic acid, bis-ethyl ester
InChI Formula InChI=1S/C12H14O4/c1-3-15-11(13)9-5-7-10(8-6-9)12(14)16-4-2/h5-8H,3-4H2,1-2H3
SMILES CCOC(=O)c1ccc(cc1)C(=O)OCC
IUPAC Name diethyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 222.2372
Formula Weight 222.0892
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.