missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Succinate, TRC

Product Code. 30382875
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30382875

Brand: Toronto Research Chemicals TRCD4450605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Succinic acid diethyl ester
CAS 123-25-1
Molecular Formula C8 H14 O4
Quantity 5 g
Synonym Succinic acid-diethyl ester, Butanedioic acid, 1,4-diethyl ester (ACI), Butanedioic acid, diethyl ester (9CI), Succinic acid, diethyl ester (6CI, 8CI), 1,4-Diethyl butanedioate, Butandioic acid diethyl ester, Clorius, DESU, Diethyl 1,2-ethanedicarboxylate, Diethyl butanedioate, Diethyl succinate, Ethyl succinate, NSC 8875
InChI Formula InChI=1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3
SMILES CCOC(=O)CCC(=O)OCC
IUPAC Name diethyl butanedioate
Molecular Weight (g/mol) 174.19
Formula Weight 174.0892
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.