missing translation for 'onlineSavingsMsg'
Learn More

Diethyl succinate, 98%

Product Code. 15189183 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Glass Bottle
Plastic Can
Plastic drum
Unit Size:
1kg
25kg
5kg
This item is not returnable. View return policy

Product Code. 15189183

Brand: Thermo Scientific Acros 150540250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-25-1 | C8H14O4 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-25-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00009208
InChI Key DKMROQRQHGEIOW-UHFFFAOYSA-N
Synonym diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
PubChem CID 31249
IUPAC Name diethyl butanedioate
SMILES CCOC(=O)CCC(=O)OCC

Specifications

Acidity 0.2mg KOH/g max.
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 217.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic Can
Linear Formula C2H5O2CCH2CH2CO2C2H5
Refractive Index 1.4190 to 1.4210
Quantity 25 kg
Beilstein 02,609
Merck Index 15,8999
Specific Gravity 1.04
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: miscible with alcohol and ether
Formula Weight 174.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Diethyl succinate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.