missing translation for 'onlineSavingsMsg'
Learn More

Diethyl succinate, 98%

Product Code. 11334499
Change view
Click to view available options
Quantity:
10,000 g
100 g
500 g
Unit Size:
10000g
100g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334499 100 g 100g
11452438 500 g 500g
11462438 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334499 Supplier Thermo Scientific Alfa Aesar Supplier No. A15777.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-25-1 | C8H14O4 | 174.196 g/mol

It is employed as emollients, masking agents, plasticisers and solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as emollients, masking agents, plasticisers and solvents.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-25-1
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00009208
InChI Key DKMROQRQHGEIOW-UHFFFAOYSA-N
Synonym diethyl succinate, ethyl succinate, butanedioic acid, diethyl ester, 1,4-diethyl butanedioate, succinic acid, diethyl ester, succinic acid diethyl ester, diethyl ethanedicarboxylate, unii-elp55c13dr, diethylester kyseliny jantarove, fema no. 2377
PubChem CID 31249
IUPAC Name diethyl butanedioate
SMILES CCOC(=O)CCC(=O)OCC

Specifications

Melting Point -21°C
Density 1.043
Boiling Point 216°C to 218°C
Flash Point 90°C (194°F)
Refractive Index 1.42
Quantity 100 g
Beilstein 907645
Merck Index 14,8869
Solubility Information Slightly soluble in water.
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material Diethyl succinate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.