missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Stilbestrol beta-D-Glucuronide, TRC

Product Code. 30008606
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008606 1 mg 1mg
30004506 5 mg 5mg
30008318 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008606 Supplier Toronto Research Chemicals Supplier No. TRCD4450201MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2408-40-4
Molecular Formula C24 H28 O8
Molecular Weight (g/mol) 444.47
Synonym Diethyl Stilbestrol-B-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[(E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenoxy]oxane-2-carboxylic acid
SMILES CC\C(=C(\CC)/c1ccc(O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O)cc1)\c3ccc(O)cc3

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C24H28O8/c1-3-17(13-5-9-15(25)10-6-13)18(4-2)14-7-11-16(12-8-14)31-24-21(28)19(26)20(27)22(32-24)23(29)30/h5-12,19-22,24-28H,3-4H2,1-2H3,(H,29,30)/b18-17+/t19-,20-,21+,22-,24+/m0/s1
Formula Weight 444.1784
Percent Purity >95%
Chemical Name or Material Diethylstilbestrol beta-D-Glucuronide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.