missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Rivastigmine, TRC

Product Code. 30378228
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378228 100 mg
30371429 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378228 Supplier Toronto Research Chemicals Supplier No. TRCD444950100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1230021-34-7
Molecular Formula C15 H24 N2 O2
Molecular Weight (g/mol) 264.3633
Synonym Carbamic acid, N,N-diethyl-, 3-[(1S)-1-(dimethylamino)ethyl]phenyl ester
IUPAC Name [3-[(1S)-1-(dimethylamino)ethyl]phenyl] N,N-diethylcarbamate
SMILES CCN(CC)C(=O)Oc1cccc(c1)[C@H](C)N(C)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H24N2O2/c1-6-17(7-2)15(18)19-14-10-8-9-13(11-14)12(3)16(4)5/h8-12H,6-7H2,1-5H3/t12-/m0/s1
Formula Weight 264.1838
Chemical Name or Material 3-[(1S)-1-(Dimethylamino)ethyl]phenyl Diethylcarbamate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.