missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, MP Biomedicals

Product Code. 11496662 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496662 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496662 Supplier MP Biomedicals Supplier No. 0215090225
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ribonuclease inhibitor

Specifications

CAS 1609-47-8
Density Density 1.12g/mL,1.1 sp. gr.
Molecular Formula C6H10O5
MDL Number MFCD00009106
Quantity 25 mL
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
SMILES CCOC(=O)OC(=O)OCC
IUPAC Name ethoxycarbonyl ethyl carbonate
Molecular Weight (g/mol) 162.14
PubChem CID 3051
ChEBI CHEBI:59051
Formula Weight 162.14
Physical Form Liquid
Chemical Name or Material Diethyl Pyrocarbonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl Pyrocarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.