missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Pyrocarbonate, >97%, Ultrapure

Product Code. 15815388 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Plastic bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15815388 100 mL Plastic bottle 100mL
15805388 25 mL Plastic bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15815388 Supplier Thermo Scientific Supplier No. J14710.AE
  • ONLINE EXCLUSIVE PRICE
    £413.10
    £339.65 / 100mL
    Save £73.45
    17% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1609-47-8 | C6H10O5 | 162.141 g/mol

  • A strong, nonspecific inhibitor of ribonuclease
  • Reacts with the N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts with amines and thus cannot be used with Tris Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Strong, nonspecific inhibitor of ribonuclease
  • Reacts with N1 of histidine residues in proteins under mild conditions
  • Used to cross-link and modify proteins by ethoxyformylation of NH2 and imidazole groups in proteins
  • Reacts
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.141
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name ethoxycarbonyl ethyl carbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Melting Point 69°C
Density 1.101
Boiling Point 93°C to 94°C (18 mmHg)
Flash Point 69°C
Packaging Plastic bottle
Refractive Index 1.398
Quantity 100 mL
Beilstein 637031
Formula Weight 162.14
Percent Purity ≥97%
Grade Ultrapure
Physical Form Liquid
Chemical Name or Material Diethyl Pyrocarbonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl pyrocarbonate, Ultrapure, Thermo Scientific
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Diethyl pyrocarbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.