missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrocarbonate, 97%, pure

Product Code. 10052140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10052140 5 g Glass bottle 5g
10417970 25 g Glass bottle 25g
10114380 100 g Glass bottle 100g
10794264 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10052140 Supplier Thermo Scientific Acros Supplier No. 170250050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1609-47-8 | C6H10O5 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-47-8
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009106
InChI Key FFYPMLJYZAEMQB-UHFFFAOYSA-N
Synonym diethyl pyrocarbonate, diethyl dicarbonate, depc, diethyl oxydiformate, baycovin, diethylpyrocarbonate, ethyl pyrocarbonate, dicarbonic acid, diethyl ester, dicarbonic acid diethyl ester, diethyl pyrocarbonic acid
PubChem CID 3051
ChEBI CHEBI:59051
IUPAC Name ethoxycarbonyl ethyl carbonate
SMILES CCOC(=O)OC(=O)OCC

Specifications

Density 1.1175g/mL
Boiling Point 93.0°C to 94.0°C (18.0 mmHg)
Flash Point 69°C
Assay Percent Range 0.97
Packaging Glass bottle
Linear Formula O(CO2C2H5)2
Refractive Index 1.3960 to 1.4000
Quantity 5 g
Beilstein 03,IV,18
Merck Index 153146
Specific Gravity 1.1175
Solubility Information Solubility in water: slow decomposition. Other solubilities: very soluble in acetone,ethanol,ligroin
Viscosity 1.97 mPa.s (20°C)
Formula Weight 162.14
Percent Purity 97%
Grade Pure
Chemical Name or Material Diethyl pyrocarbonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl pyrocarbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.