missing translation for 'onlineSavingsMsg'
Learn More

Diethyl pyrazole-3,5-dicarboxylate, 99%, Thermo Scientific Chemicals

Product Code. p-109830 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10166683

Brand: Thermo Scientific Acros 296520010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37687-24-4
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.21
MDL Number MFCD00152167
InChI Key MBWXLICVQZUJOW-UHFFFAOYSA-N
Synonym diethyl 3,5-pyrazoledicarboxylate, diethyl pyrazole-3,5-dicarboxylate, 3,5-diethoxycarbonylpyrazole, 1h-pyrazole-3,5-dicarboxylic acid diethyl ester, 3,5-pyrazoledicarboxylic acid diethyl ester, 3,5-diethyl 1h-pyrazole-3,5-dicarboxylate, 1h-pyrazole-3,5-dicarboxylic acid, diethyl ester, zlchem 861, pubchem22955, acmc-1aj76
PubChem CID 142184
IUPAC Name diethyl 1H-pyrazole-3,5-dicarboxylate
SMILES CCOC(=O)C1=CC(=NN1)C(=O)OCC

Specifications

Melting Point 51.0°C to 55.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 212.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Diethyl pyrazole-3,5-dicarboxylate, 99%
compliance-icons
Product Identifier
  • Diethyl pyrazole-3,5-dicarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.