missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Phthalate (Phenyl-13C6, D4), TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Phthalate (Phenyl-13C6, D4) |
| Molecular Formula | C17 H13 Cl N4 O . C H4 . H2 O |
| Quantity | 5 mg |
| Synonym | 1,2-Benzenedicarboxylic Acid 1,2-Diethyl Ester (Phenyl-13C6, D4), 1,2-Benzenedicarboxylic Acid Diethyl Ester (Phenyl-13C6, D4), Phthalic Acid Diethyl Ester (Phenyl-13C6, D4), Anozol (Phenyl-13C6, D4), DEP (Phenyl-13C6, D4), Diethyl 1,2-Benzenedicarboxylate (Phenyl-13C6, D4), Diethyl Phthalate (Phenyl-13C6, D4), Ethyl Phthalate (Phenyl-13C6, D4), NSC 8905 (Phenyl-13C6, D4), Neantine (Phenyl-13C6, D4), Palatinol A (Phenyl-13C6, D4), Phthalol (Phenyl-13C6, D4), Placidol E (Phenyl-13C6, D4), Solvanol (Phenyl-13C6, D4), Unimoll DA (Phenyl-13C6, D4), o-Benzenedicarboxylic Acid Diethyl Ester (Phenyl-13C6, D4), o-Bis(ethoxycarbonyl)benzene (Phenyl-13C6, D4) |
| InChI Formula | InChI=1S/C17H13ClN4O.CH4.H2O/c1-10-20-21-16-17(23)19-15(11-5-3-2-4-6-11)13-9-12(18)7-8-14(13)22(10)16;;/h2-9,17,23H,1H3;1H4;1H2 |
| SMILES | C.O.Cc1nnc2C(O)N=C(c3ccccc3)c4cc(Cl)ccc4n12 |
| IUPAC Name | 8-chloro-1-methyl-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepin-4-ol;methane;hydrate |
| Molecular Weight (g/mol) | 358.822 |
| Formula Weight | 358.12 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?