missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Phthalate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-ethyl ester D4 |
| CAS | 93952-12-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 2H4 H10 O4 |
| Quantity | 100 mg |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, diethyl ester (9CI), Phthalic acid diethyl ester (3,4,5,6) D4, Phthalic acid, bis-ethyl ester D4, Diethyl phthalate-d4, Phthalic Acid, Diethyl Ester D4 (phenyl D4) |
| InChI Formula | InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i5D,6D,7D,8D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OCC)c(C(=O)OCC)c1[2H] |
| IUPAC Name | diethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 226.26 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?