missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Phthalate-d14, TRC

Product Code. 30387634
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387634 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30387634 Supplier Toronto Research Chemicals Supplier No. TRCD44477450MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phthalic acid diethyl ester d14
CAS 99873-99-1
Molecular Formula C12 2H14 O4
Quantity 50 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, di(ethyl-d5) ester (9CI), Phthalic acid diethyl ester D14, Phthalic acid, bis-ethyl ester D14, Diethyl phthalate-d14
InChI Formula InChI=1S/C12H14O4/c1-3-15-11(13)9-7-5-6-8-10(9)12(14)16-4-2/h5-8H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2,5D,6D,7D,8D
SMILES [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])C([2H])([2H])[2H])c(C(=O)OC([2H])([2H])C([2H])([2H])[2H])c1[2H]
IUPAC Name bis(1,1,2,2,2-pentadeuterioethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molecular Weight (g/mol) 236.32
Formula Weight 236.1771
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.