missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phthalate, 99%

Product Code. 11408157
Change view
Click to view available options
Quantity:
250 g
1000 g
2500 g
Unit Size:
1000g
2500g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408157 1000 g 1000g
11418157 250 g 250g
11428157 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408157 Supplier Thermo Scientific Alfa Aesar Supplier No. A17529.0B
  • ONLINE EXCLUSIVE PRICE
    £35.09
    £28.65 / 1000g
    Save £6.44
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 84-66-2 | C12H14O4 | 222.24 g/mol

Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-66-2
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009111
InChI Key FLKPEMZONWLCSK-UHFFFAOYSA-N
Synonym diethyl phthalate, phthalic acid diethyl ester, ethyl phthalate, anozol, neantine, phthalol, solvanol, diethyl o-phthalate, palatinol a, placidol e
PubChem CID 6781
ChEBI CHEBI:34698
IUPAC Name diethyl benzene-1,2-dicarboxylate
SMILES CCOC(=O)C1=CC=CC=C1C(=O)OCC

Specifications

Melting Point -64°C
Density 1.118
Boiling Point 296°C to 298°C
Flash Point 160°C (320°F)
Refractive Index 1.502
Quantity 1000 g
Beilstein 1912500
Merck Index 14,7371
Solubility Information Miscible with ethanol,chloroform,diethyl ether,benzene,acetone,vegetable oils,ketones,carbon tetrachloride,esters,aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material Diethyl phthalate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl phthalate
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.