missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphate, MedChemExpress

Product Code. p-200096993
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16491526

Brand: Medchem Express HY101417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos.

Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos.

Specifications

Chemical Name or Material Diethyl phosphate
Physical Form Liquid
CAS 598-02-7
Color Colorless
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C4H11O4P
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Diethyl phosphoric acid
Solubility Information DMSO : ≥ 34 mg/mL (220.64 mM)
SMILES O=P(OCC)(O)OCC
Molecular Weight (g/mol) 154.1
Formula Weight 154.1
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.