missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phosphate, MedChemExpress

Product Code. 16491526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491526 500 mg 500mg
16484596 100 mg 100mg
16480926 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491526 Supplier Medchem Express Supplier No. HY101417
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos.

Diethylphosphate (DEP) is product of metabolism and of environmental degradation of a commonly used insecticide Chlorpyrifos.

Specifications

Chemical Name or Material Diethyl phosphate
Physical Form Liquid
CAS 598-02-7
Color Colorless
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C4H11O4P
For Use With (Application) Metabolism-protein/nucleotide metabolism
Synonym Diethyl phosphoric acid
Solubility Information DMSO : ≥ 34 mg/mL (220.64 mM)
SMILES O=P(OCC)(O)OCC
Molecular Weight (g/mol) 154.1
Formula Weight 154.1
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.