missing translation for 'onlineSavingsMsg'
Learn More

Diethyl phenylthiomethylphosphonate, 96%

Product Code. 11440254
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440254 50 g 50g
11430254 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11440254 Supplier Thermo Scientific Alfa Aesar Supplier No. A13660.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38066-16-9 | C11H17O3PS | 260.288 g/mol

Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phenylthiomethylphosphonate is used to produce Diethylphosphorylmethylphenylsulfoxid, Chemical reagents, pharmaceutical research, pharmaceutical intermediates.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38066-16-9
Molecular Formula C11H17O3PS
Molecular Weight (g/mol) 260.288
MDL Number MFCD00015130
InChI Key FBUXEPJJFVDUFE-UHFFFAOYSA-N
Synonym diethyl phenylthiomethylphosphonate, diethyl phenylthio methyl phosphonate, diethyl phenylsulfanyl methylphosphonate, phosphonic acid, phenylthio methyl-, diethyl ester, diethoxy phenylthiomethyl phosphino-1-one, diethylphenylthiomethylphosphonate, acmc-1bn85, diethyl-phenylthiomethylphosphonate, diethyl methylthiobenzyl phosphonate, diethyl phenylsulfanyl methylphosphonate #
PubChem CID 99227
IUPAC Name diethoxyphosphorylmethylsulfanylbenzene
SMILES CCOP(=O)(CSC1=CC=CC=C1)OCC

Specifications

Density 1.174
Boiling Point 146°C to 148°C (0.5 mmHg)
Refractive Index 1.534
Quantity 50 g
Beilstein 1246215
Solubility Information Difficult to mix.
Formula Weight 260.29
Percent Purity 96%
Chemical Name or Material Diethyl phenylthiomethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.