missing translation for 'onlineSavingsMsg'
Learn More

Diethyl p-Toluenesulfonyloxymethylphosphonate, TRC

Product Code. 30378227
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378227 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378227 Supplier Toronto Research Chemicals Supplier No. TRCD44526010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl p-Toluenesulfonyloxymethylphosphonate
CAS 31618-90-3
Molecular Formula C12 H19 O6 P S
Quantity 10 g
Synonym Phosphonic acid, P-[[[(4-methylphenyl)sulfonyl]oxy]methyl]-, diethyl ester (ACI), Diethyl P-[[[(4-methylphenyl)sulfonyl]oxy]methyl]phosphonate (ACI), Phosphonic acid, (hydroxymethyl)-, diethyl ester, p-toluenesulfonate (8CI), Phosphonic acid, [[[(4-methylphenyl)sulfonyl]oxy]methyl]-, diethyl ester (9CI), (Diethoxyphosphoryl)methyl 4-methylbenzenesulfonate, 4-Methylbenzenesulfonic acid (diethoxyphosphoryl)methyl ester, [[[(4-Tolyl)sulfonyl]oxy]methyl]phosphonic acid diethyl ester, Diethoxyphosphorylmethyl tosylate, Diethyl (tosyloxy)methylphosphonate, Diethyl [[(p-toluenesulfonyl)oxy]methyl]phosphonate, Diethyl [[[(4-tolyl)sulfonyl]oxy]methyl]phosphonate, Diethyl p-tosyloxymethylphosphonate, Tosyloxymethyl diethyl phosphonate
InChI Formula InChI=1S/C12H19O6PS/c1-4-16-19(13,17-5-2)10-18-20(14,15)12-8-6-11(3)7-9-12/h6-9H,4-5,10H2,1-3H3
SMILES CCOP(=O)(COS(=O)(=O)c1ccc(C)cc1)OCC
IUPAC Name diethoxyphosphorylmethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 322.31
Formula Weight 322.064
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.