missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Oxalacetate Sodium Salt, TRC

Product Code. 30381188
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381188 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381188 Supplier Toronto Research Chemicals Supplier No. TRCD4446955G
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Oxalacetate Sodium Salt
CAS 40876-98-0
Molecular Formula C8H11NaO5
Quantity 5 g
Synonym 2-(Oxo)succinic Acid Diethyl Ester Monosodium Salt, Diethyl 2-Hydroxy-2-butenedioate Sodium Salt, Diethyl Oxobutanedioate Sodium Salt, Diethyl Sodiooxalacetate, Diethyl Sodium Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt, Sodium 1,4-Diethoxy-1,4-dioxobut-2-en-2-olate, Sodium Diethyl Oxalacetate, Sodium Diethyl Oxaloacetate, Sodium Diethyl Oxobutanedioate
InChI Formula InChI=1S/C8H11O5.Na/c1-3-12-7(10)5-6(9)8(11)13-4-2;/h5H,3-4H2,1-2H3;/q-1;+1
SMILES [Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
IUPAC Name sodium;diethyl 2-oxobutanedioate
Molecular Weight (g/mol) 210.16
Formula Weight 210.0504
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.