Learn More
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Color | Beige to Orange |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% |
| Packaging | Plastic bottle |
| Linear Formula | C2H5OCOCH=C(ONa)COOC2H5 |
| Quantity | 500 g |
| Merck Index | 14, 3836 |
| Formula Weight | 210.16 |
| Percent Purity | 95% |
| Grade | Practical |
| Vedi altri risultati |
Sicurezza e movimentazione
- Diethyl oxalacetate sodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.