missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-propylmalonate, 99%

Product Code. 11470907
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470907 25 g 25g
11480907 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470907 Supplier Thermo Scientific Alfa Aesar Supplier No. B23304.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2163-48-6 | C10H18O4 | 202.25 g/mol

Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl n-propylmalonate was used in the synthesis of propylacrylic acid (PAA).

Solubility
Soluble in Dichloromethane, Ethanol and Ethyl Acetate.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2163-48-6
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00009168
InChI Key GRRSDGHTSMJICM-UHFFFAOYSA-N
Synonym diethyl propylmalonate, diethyl 2-propylmalonate, 1,3-diethyl 2-propylpropanedioate, diethyl n-propylmalonate, diethylpropylmalonate, propylmalonic acid diethyl ester, propanedioic acid, propyl-, diethyl ester, diethyl-n-propylmalonate, malonic acid, propyl-, diethyl ester, diethyl propyl malonate
PubChem CID 16552
IUPAC Name diethyl 2-propylpropanedioate
SMILES CCCC(C(=O)OCC)C(=O)OCC

Specifications

Density 0.987
Boiling Point 221°C to 222°C
Flash Point 91°C (197°F)
Refractive Index 1.4205
Quantity 25 g
Solubility Information Soluble in Dichloromethane,Ethanol and Ethyl Acetate.
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Diethyl n-propylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.