missing translation for 'onlineSavingsMsg'
Learn More

Diethyl n-dodecylphosphonate, ≥97%, Thermo Scientific Chemicals

Product Code. 15437807
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15437807

Brand: Thermo Scientific Alfa Aesar H37697.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4844-38-6
Molecular Formula C16H35O3P
Molecular Weight (g/mol) 306.427
MDL Number MFCD00015595
InChI Key LRIWHHSVZCKNJL-UHFFFAOYSA-N
Synonym diethyl dodecylphosphonate, phosphonic acid, dodecyl-, diethyl ester, diethyl 1-dodecyl phosphonate, phosphonic acid, p-dodecyl-, diethyl ester, dodecylphosphonic acid diethyl ester, phosphonic acid,p-dodecyl-, diethyl ester
PubChem CID 78558
IUPAC Name 1-diethoxyphosphoryldodecane
SMILES CCCCCCCCCCCCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 168°C (1.5 mmHg)
Quantity 1 g
Formula Weight 306.42
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material Diethyl n-dodecylphosphonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.