missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Methylphenylmalonate, TRC

Product Code. 30387433
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387433 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387433 Supplier Toronto Research Chemicals Supplier No. TRCD473613250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Methylphenylmalonate
CAS 34009-61-5
Molecular Formula C4 H4 Cl N S
Quantity 250 mg
Synonym 2-Methyl-2-phenyl-propanedioic Acid 1,3-Diethyl Ester, Methylphenyl-malonic Acid Diethyl Ester, Methylphenyl-propanedioic Acid Diethyl Ester, Diethyl 2-Methyl-2-phenylmalonate, NSC 147481
InChI Formula InChI=1S/C4H4ClNS/c5-3-4-6-1-2-7-4/h1-2H,3H2
SMILES ClCc1nccs1
IUPAC Name 2-(chloromethyl)-1,3-thiazole
Molecular Weight (g/mol) 133.599
Formula Weight 132.975
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.