missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Methylphenylmalonate, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Methylphenylmalonate |
| CAS | 34009-61-5 |
| Molecular Formula | C4 H4 Cl N S |
| Quantity | 100 mg |
| Synonym | 2-Methyl-2-phenyl-propanedioic Acid 1,3-Diethyl Ester, Methylphenyl-malonic Acid Diethyl Ester, Methylphenyl-propanedioic Acid Diethyl Ester, Diethyl 2-Methyl-2-phenylmalonate, NSC 147481 |
| InChI Formula | InChI=1S/C4H4ClNS/c5-3-4-6-1-2-7-4/h1-2H,3H2 |
| SMILES | ClCc1nccs1 |
| IUPAC Name | 2-(chloromethyl)-1,3-thiazole |
| Molecular Weight (g/mol) | 133.599 |
| Formula Weight | 132.975 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?