missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Malonate-13C3, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl Malonate-13C3 |
| CAS | 53051-81-3 |
| Molecular Formula | C413C3H12O4 |
| Quantity | 250 mg |
| Synonym | Propanedioic Acid 1,3-Diethyl Ester-13C3, Malonic Acid Diethyl Ester-13C3, Propanedioic Acid Diethyl Ester-13C3, Carbethoxyacetic Ester-13C3, Dicarbethoxymethane-13C3, Diethyl Propane-1,3-dioate-13C3, Diethyl Propanedioate-13C3, Ethyl Malonate-13C3, Malonate Diethyl Ester-13C3, Methanedicarboxylic Acid Diethyl Ester-13C3, NSC 136903-13C3, NSC 8864-13C3 |
| InChI Formula | InChI=1S/C7H12O4/c1-3-10-6(8)5-7(9)11-4-2/h3-5H2,1-2H3/i5+1,6+1,7+1 |
| SMILES | O=[13C]([13C]([H])([H])[13C](OCC)=O)OCC |
| Molecular Weight (g/mol) | 163.15 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?