missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Maleate, TRC

Product Code. 30137365
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137365 25 g 25g
17961715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137365 Supplier Toronto Research Chemicals Supplier No. TRCD44453525G
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-05-9
Molecular Formula C8 H12 O4
Molecular Weight (g/mol) 172.18
Synonym 2-Butenedioic acid (2Z)-, diethyl ester (9CI), 2-Butenedioic acid (Z)-, diethyl ester, Maleic acid, diethyl ester (6CI,8CI), (2Z)-2-Butenedioic acid diethyl ester, (Z)-2-Butenedioic acid diethyl ester, Diethyl (Z)-2-butenedioate, Diethyl maleate, Ethyl maleate, Maleinic acid diethyl ester, Staflex DEM, Diethyl Maleate
IUPAC Name diethyl (Z)-but-2-enedioate
SMILES CCOC(=O)\C=C/C(=O)OCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H12O4/c1-3-11-7(9)5-6-8(10)12-4-2/h5-6H,3-4H2,1-2H3/b6-5-
Formula Weight 172.0736
Chemical Name or Material Diethyl Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.