missing translation for 'onlineSavingsMsg'
Learn More

Diethyl maleate, 97%

Product Code. 11326696
Change view
Click to view available options
Quantity:
5 g
250 g
1000 g
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326696 1000 g 1kg
11306696 5 g 5g
11316696 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326696 Supplier Thermo Scientific Alfa Aesar Supplier No. L03876.0B
  • ONLINE EXCLUSIVE PRICE
    £62.20
    £55.65 / 1kg
    Save £6.55
    10% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 141-05-9 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-05-9
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00009191
InChI Key IEPRKVQEAMIZSS-WAYWQWQTSA-N
Synonym diethyl maleate, ethyl maleate, maleic acid, diethyl ester, diethylmaleate, maleic acid diethyl ester, 2-butenedioic acid z-, diethyl ester, diethyl z-but-2-enedioate, diethyl 2z-but-2-enedioate, unii-g81wqb56ol, ccris 941
PubChem CID 5271566
ChEBI CHEBI:68508
IUPAC Name diethyl (Z)-but-2-enedioate
SMILES CCOC(=O)C=CC(=O)OCC

Specifications

Melting Point -11°C
Density 1.064
Boiling Point 221°C to 224°C
Flash Point 93°C (199°F)
Odor Pleasant
Refractive Index 1.44
Quantity 1000 g
Beilstein 1100825
Merck Index 14,3123
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Diethyl maleate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.