missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ketomalonate, 95%

Product Code. 10366320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10366320 25 g Glass bottle 25g
10253950 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366320 Supplier Thermo Scientific Supplier No. 114430250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 609-09-6 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-09-6
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00009121
InChI Key DBKKFIIYQGGHJO-UHFFFAOYSA-N
Synonym diethyl ketomalonate, diethyl mesoxalate, diethyl 2-oxomalonate, diethyl oxomalonate, ethyl mesoxalate, ethyl ketomalonate, propanedioic acid, oxo-, diethyl ester, diethylketomalonate, mesoxalic acid diethyl ester, mesoxalic acid, diethyl ester
PubChem CID 69105
IUPAC Name diethyl 2-oxopropanedioate
SMILES CCOC(=O)C(=O)C(=O)OCC

Specifications

Color Yellow
Density 1.1420g/mL
Boiling Point 208.0°C to 210.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O2CCOCO2C2H5
Refractive Index 1.4150 to 1.4190
Quantity 25 g
Beilstein 03, 769
Fieser 06,188; 08,170
Merck Index 15, 5983
Specific Gravity 1.142
Formula Weight 174.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Diethyl ketomalonate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl ketomalonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.