missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isopropylmalonate, 98+%

Product Code. 11437066
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11437066 50 g 50g
11427066 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11437066 Supplier Thermo Scientific Alfa Aesar Supplier No. B25384.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 759-36-4 | C10H18O4 | 202.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 759-36-4
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00040491
InChI Key BYQFBFWERHXONI-UHFFFAOYSA-N
Synonym diethyl isopropylmalonate, diethyl 2-isopropylmalonate, isopropyl diethyl malonate, 1,3-diethyl 2-propan-2-yl propanedioate, diethylisopropylmalonate, isopropylmalonic acid diethyl ester, propanedioic acid, 1-methylethyl-, diethyl ester, ethyl isopropylmalonate, 1,3-diethyl 2-isopropylpropanedioate, 2-isopropyl-malonic acid diethyl ester
PubChem CID 12966
IUPAC Name diethyl 2-propan-2-ylpropanedioate
SMILES CCOC(=O)C(C(C)C)C(=O)OCC

Specifications

Density 0.981
Boiling Point 102°C to 104°C (12 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.421
Quantity 50 g
Beilstein 1101711
Formula Weight 202.25
Percent Purity ≥98%
Chemical Name or Material Diethyl isopropylmalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.