missing translation for 'onlineSavingsMsg'
Learn More

Diethyl isopropylidenemalonate, 97%

Product Code. 15284744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15284744 5 g 5g
15294744 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284744 Supplier Thermo Scientific Acros Supplier No. 443310050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6802-75-1 | C10H16O4 | 200.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6802-75-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.23
InChI Key WEISAZNMMVPNTH-UHFFFAOYSA-N
Synonym diethyl isopropylidenemalonate, diethyl 2-propan-2-ylidene malonate, diethylisopropylidenemalonate, malonic acid, isopropylidene-, diethyl ester, propanedioic acid, 1-methylethylidene-, diethyl ester, isopropylidenemalonic acid diethyl ester, 1,3-diethyl 2-propan-2-ylidene propanedioate, diethyl 2-1-methylethylidene malonate, 2-isopropylidene-malonic acid diethyl ester, acmc-209o1m
PubChem CID 81255
IUPAC Name diethyl 2-propan-2-ylidenepropanedioate
SMILES CCOC(=O)C(=C(C)C)C(=O)OCC

Specifications

Density 1.0210g/mL
Boiling Point 175°C to 178°C (120.0 mmHg)
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.021
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Diethyl isopropylidenemalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.