missing translation for 'onlineSavingsMsg'
Learn More

Diethyl iminodicarboxylate, 98%

Product Code. 15587529 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15587529 5 g 5g
15577529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15587529 Supplier Thermo Scientific Acros Supplier No. 460770050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19617-44-8 | C6H11NO4 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19617-44-8
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
InChI Key PQVSTLUFSYVLTO-UHFFFAOYSA-N
Synonym azamalonic ester, diethyl imidodicarbonate, diethyl azamalonate, imidodicarbonic acid, diethyl ester, diethyl iminodicarboxylate, diethylazamalonate, imidodicarbonic acid,1,3-diethyl ester, diethylimidodicarbonate, diethyl imidodicarbonate #, ethyl ethoxycarbonylamino formate
PubChem CID 140564
IUPAC Name ethyl N-ethoxycarbonylcarbamate
SMILES CCOC(=O)NC(=O)OCC

Specifications

Melting Point 51.0°C to 53.0°C
Boiling Point 132.0°C to 134.0°C (15.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Formula Weight 161.16
Percent Purity 98%
Chemical Name or Material Diethyl iminodicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.