missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Iminodiacetate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Diethyl Iminodiacetate |
| CAS | 6290-05-7 |
| Molecular Formula | C8H15NO4 |
| Quantity | 1 g |
| Synonym | Iminodiacetic Acid Diethyl Ester, Iminodiethyl Diacetate, N,N-Bis(ethoxycarbonylmethyl)amine, N-(Ethoxycarbonylmethyl)glycine Ethyl Ester, NSC 6950, [[(Ethoxycarbonyl)methyl]amino]acetic Acid Ethyl Ester, 66: PN: US20050107325 PAGE: 155 claimed protein, Diethyl Iminobis(acetate) |
| InChI Formula | InChI=1S/C8H15NO4/c1-3-12-7(10)5-9-6-8(11)13-4-2/h9H,3-6H2,1-2H3 |
| SMILES | CCOC(=O)CNCC(=O)OCC |
| IUPAC Name | ethyl 2-[(2-ethoxy-2-oxoethyl)amino]acetate |
| Molecular Weight (g/mol) | 189.21 |
| Formula Weight | 189.1001 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction