missing translation for 'onlineSavingsMsg'
Learn More

Diethyl hydroxymethylphosphonate, tech. 85%, Thermo Scientific Chemicals

Product Code. 11424727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424727 25 g 25g
11414727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424727 Supplier Thermo Scientific Alfa Aesar Supplier No. B21215.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3084-40-0 | Molecular Formula: C5H13O4P | Molecular Weight: 168.129 g/mol

Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl hydroxymethylphosphonate acts as a reactant for synthesis of α And β-dialkoxyphosphoryl isothiocyanate used in antiproliferative studies, dendritic polyglycerol anions for L-selectin inhibition, phosphonates for anti-HIV-1 activity, homo and copolymers of phosphonated norbornenes, Mitsunobu reactions. It is involved in studies of FBPase inhibition by phosphonic acid-containing thaizoles.

Notes
Incompatible with oxidizing agents. Store in a cool, dry condition in a well sealed container.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3084-40-0
Molecular Formula C5H13O4P
Molecular Weight (g/mol) 168.129
MDL Number MFCD00014418
InChI Key RWIGWWBLTJLKMK-UHFFFAOYSA-N
Synonym diethyl hydroxymethyl phosphonate, diethyl hydroxymethylphosphonate, phosphonic acid, hydroxymethyl-, diethyl ester, hydroxymethyl phosphonic acid, diethyl ester, hydroxymethyl phosphonic acid diethyl ester, phosphonic acid, p-hydroxymethyl-, diethyl ester, acmc-1corl, diethoxyphosphoryl-methanol
PubChem CID 76513
IUPAC Name diethoxyphosphorylmethanol
SMILES CCOP(=O)(CO)OCC

Specifications

Density 1.14
Boiling Point 124°C to 126°C (3 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.444
Quantity 25 g
Formula Weight 168.13
Percent Purity 85%
Grade Technical
Chemical Name or Material Diethyl hydroxymethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.