missing translation for 'onlineSavingsMsg'
Learn More

Diethyl fumarate, 98%

Product Code. 11453307
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453307 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453307 Supplier Thermo Scientific Alfa Aesar Supplier No. A16214.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 623-91-6 | C8H12O4 | 172.18 g/mol

Diethyl fumarate, is used as an intermediate for plasticizer, flavoring agents and surface coatings. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a intermediate and solvent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl fumarate, is used as an intermediate for plasticizer, flavoring agents and surface coatings. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a intermediate and solvent in organic synthesis.

Solubility
It is soluble in water, acetone, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases, reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-91-6
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00064455
InChI Key IEPRKVQEAMIZSS-AATRIKPKSA-N
Synonym diethyl fumarate, fumaric acid, diethyl ester, anti-psoriaticum, diethyl 2e-but-2-enedioate, fumaric acid diethyl ester, diethyl e-but-2-enedioate, 2-butenedioic acid e-, diethyl ester, trans-2-butenedioic acid diethyl ester, 2-butenedioic acid 2e-, diethyl ester, unii-5wbu5a3e8a
PubChem CID 638144
ChEBI CHEBI:87388
IUPAC Name diethyl (E)-but-2-enedioate
SMILES CCOC(=O)\C=C\C(=O)OCC

Specifications

Melting Point ∼0°C
Density 1.05
Boiling Point 98°C to 100°C (14 mmHg)
Flash Point 91°C (195°F)
Refractive Index 1.44
Quantity 500 g
Beilstein 775347
Solubility Information It is soluble in water,acetone,and chloroform.
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material Diethyl fumarate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.