missing translation for 'onlineSavingsMsg'
Learn More

Diethyl fluoromalonate, 97%, Thermo Scientific™

Product Code. 10227230 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 10227230

Brand: Thermo Scientific Acros 307640010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 685-88-1
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 121°C to 122°C (30.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H11FO4
Linear Formula FCH(CO2C2H5)2
MDL Number MFCD00009139
Quantity 1 mL
Specific Gravity 1.12
Synonym diethyl fluoromalonate, diethyl 2-fluoromalonate, fluoromalonic acid diethyl ester, 1,3-diethyl 2-fluoropropanedioate, diethylfluoromalonate, 2-fluoro-malonic acid diethyl ester, diethyl2-fluoromalonate, diethyl fluoropropanedioate, propanedioic acid, fluoro-, diethyl ester, malonic acid, fluoro-, diethyl ester
InChI Key GOWQBFVDZPZZFA-UHFFFAOYSA-N
SMILES CCOC(=O)C(C(=O)OCC)F
IUPAC Name diethyl 2-fluoropropanedioate
Molecular Weight (g/mol) 178.16
PubChem CID 12702
Formula Weight 178.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Diethyl fluoromalonate, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.