missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl ethylmalonate, 99%

Product Code. 11401077
Change view
Click to view available options
Quantity:
100g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401077 100g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11401077 Supplier Alfa Aesar™ Supplier No. B24077.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 133-13-1
Boiling Point 204°C to 206°C
Flash Point 88°C (190°F)
Molecular Formula C9H16O4
MDL Number MFCD00009167
Quantity 100g
Beilstein 607635
Synonym diethyl ethylmalonate, diethyl 2-ethylmalonate, propanedioic acid, ethyl-, diethyl ester, ethylmalonic acid diethyl ester, 1,3-diethyl 2-ethylpropanedioate, unii-69g92oyr3m, propanedioic acid, 2-ethyl-, 1,3-diethyl ester, malonic acid, ethyl-, diethyl ester, diethyl ethylpropanedioate, diethyl 2-ethylpropane-1,3-dioate
InChI Key VQAZCUCWHIIFGE-UHFFFAOYSA-N
SMILES CCC(C(=O)OCC)C(=O)OCC
IUPAC Name diethyl 2-ethylpropanedioate
Molecular Weight (g/mol) 188.223
PubChem CID 8610
Formula Weight 188.22
Percent Purity 99%
Chemical Name or Material Diethyl ethylmalonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.