missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ethylidenemalonate, 99%

Product Code. 11328625
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328625 5 g 5g
11318625 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328625 Supplier Thermo Scientific Alfa Aesar Supplier No. L12117.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1462-12-0 | C9H14O4 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1462-12-0
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00009145
InChI Key LBBAWVLUOZVYCC-UHFFFAOYSA-N
Synonym diethyl ethylidenemalonate, diethyl 2-ethylidenemalonate, propanedioic acid, ethylidene-, diethyl ester, 1,3-diethyl 2-ethylidenepropanedioate, 2-ethylidenemalonic acid diethyl ester, diethylethylidenemalonate, acmc-1c0mi, diethyl ethylidene malonate, diethyl ethylidenepropanedioate, diethyl 2-ethylidenemalonate #
PubChem CID 73831
SMILES CCOC(=O)C(=CC)C(=O)OCC

Spezifikation

Density 1.019
Boiling Point 115°C to 116°C (17 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.442
Quantity 5 g
Beilstein 1773932
IUPAC Name 1,3-diethyl 2-ethylidenepropanedioate
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material Diethyl ethylidenemalonate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.