missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ether, Tested according to Ph.Eur., Honeywell™

Product Code. 15633510
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15633510 1 L 1 L
15643510 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15633510 Supplier Honeywell Riedel-de Haën Supplier No. 317011L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Tekniske data

Color Colorless
Density 0.706g/mL at 25°C
Quantity 1 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Linear Formula (CH3CH2)2O
Vapor Pressure 2311 hPa (60°C)
UN Number 1155
Beilstein 1696894
Formula Weight 74.12g/mol
Chemical Name or Material Diethyl Ether, Tested according to Ph.Eur.
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.