missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:
- Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
- Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
- Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation
Specifications
Specifications
| Melting Point | -116.3°C |
| CAS | 60-29-7 |
| Color | Colorless |
| Acidity | Max. 0.0005meq/g |
| Density | 0.71g/cm3 at 20°C |
| Quantity | 2.5 L |
| Explosion Limit | 1.7 - 48 Vol % |
| Flash Point | −45°C |
| Ignition Point | 170°C |
| Odor | Sweetish |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?