missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether stabilised, BAKER ANALYZED™, J.T. Baker™

Product Code. 10382931 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10382931 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10382931 Supplier JT Baker Supplier No. 8401.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point -123°C
CAS 60-29-7
Color Max. 10 (APHA)
Density 0.7135 g/cm³ (20°C)
Quantity 1 L
Flash Point -40°C
Odor Passes Test
Packaging Glass Bottle
Residue after Evaporation max. 0.001%
Assay Percent Range min. 99.5%
Molecular Formula C4H10O
MDL Number MFCD00011646
Water max. 0.01%
Boiling Point 35°C (1013 hPa)
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
SMILES CCOCC
IUPAC Name ethoxyethane
Molecular Weight (g/mol) 74.12
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.