missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Spectrophotometric Grade, 99+%, stab. with copper

Product Code. 15498575
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
4 x 1 L
Unit Size:
1 L
1000mL
2500mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498575 4 x 1 L 1 L
15478575 500 mL 500mL
15488575 1000 mL 1000mL
15468575 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15498575 Supplier Thermo Scientific Alfa Aesar Supplier No. H33678.M1
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-29-7 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym Ether, Ethyl ether
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Quantity 4 x 1 L
Boiling Point 34.6°C
Percent Purity 99+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.